1-(Perfluoro-n-octyl)hexadecane

Supplier: Matrix Scientific

009127-1G 009127-5G
101796-428EA 253.85 USD
101796-428 101796-430
1-(Perfluoro-n-octyl)hexadecane
1-(Perfluoro-n-octyl)hexadecane

Hazard: Irritant

Formula: C₂₄H₃₃F₁₇
Boiling Pt: 569 °C
Melting Pt: 108 °C
MDL Number: MFCD04038347
CAS Number: 117146-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR