1-(Perfluoro-n-octyl)dodecane

Supplier: Matrix Scientific

009126-1G 009126-5G
101796-424EA 253.85 USD
101796-424 101796-426
1-(Perfluoro-n-octyl)dodecane
1-(Perfluoro-n-octyl)dodecane

Hazard: Irritant

Formula: C₂₀H₂₅F₁₇
Boiling Pt: 479 °C
Melting Pt: 71 °C
MDL Number: MFCD04038346
CAS Number: 106873-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR