1-(Perfluoro-n-octyl)decane

Supplier: Matrix Scientific

009125-1G 009125-5G
101796-420EA 90.77 USD
101796-420 101796-422
1-(Perfluoro-n-octyl)decane
1-(Perfluoro-n-octyl)decane

Hazard: Irritant

Formula: C₁₈H₂₁F₁₇
Boiling Pt: 427 °C
Melting Pt: 52 °C
MDL Number: MFCD04038345
CAS Number: 138472-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR