Perfluorooctanamidine ≥97%

Supplier: Matrix Scientific

009118-1G 009118-5G
101796-404EA 438.46 USD
101796-404 101796-406
Perfluorooctanamidine ≥97%
Perfluorooctanamidine

Hazard: Irritant

Formula: C₈H₃F₁₅N₂
MDL Number: MFCD00069165
CAS Number: 307-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR