1-(Perfluoro-n-hexyl)tetradecane

Supplier: Matrix Scientific

009113-500MG 009113-2.5G
101796-386EA 467.69 USD
101796-386 101796-388
1-(Perfluoro-n-hexyl)tetradecane
1-(Perfluoro-n-hexyl)tetradecane

Hazard: Irritant

Formula: C₂₀H₂₉F₁₃
Boiling Pt: 481°C
Melting Pt: 67 °C
Density: 1.201
Flash Pt: 149.6°C
MDL Number: MFCD04038341
CAS Number: 154628-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR