1-(Perfluoro-n-hexyl)dodecane

Supplier: Matrix Scientific

009112-500MG 009112-2.5G
101796-382EA 100 USD
101796-382 101796-384
1-(Perfluoro-n-hexyl)dodecane
1-(Perfluoro-n-hexyl)dodecane

Hazard: Irritant

Formula: C₁₈H₂₅F₁₃
Boiling Pt: 435°C
Melting Pt: 48 °C
MDL Number: MFCD04038340
CAS Number: 90499-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR