Perfluoro(1-ethyl-3-propoxycyclohexane) ≥97%

Supplier: Matrix Scientific

009102-1G 009102-5G
101796-346EA 144.62 USD
101796-346 101796-348
Perfluoro(1-ethyl-3-propoxycyclohexane) ≥97%
Perfluoro(1-ethyl-3-propoxycyclohexane)

Hazard: Irritant

Formula: C₁₁F₂₂O
MDL Number: MFCD03094121
CAS Number: 400626-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR