1H,1H-Perfluoro-3,6-dioxadecan-1-ol ≥98%

Supplier: Matrix Scientific

009098-5G 009098-25G
101796-336EA 129.23 USD
101796-336 101796-338
1H,1H-Perfluoro-3,6-dioxadecan-1-ol ≥98%
1H,1H-Perfluoro-3,6-dioxadecan-1-ol

Hazard: Irritant

Formula: C₈H₃F₁₅O₃
MDL Number: MFCD03094403
CAS Number: 152914-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR