Perfluoro-3,6-dioxadecanoic acid ≥98%

Supplier: Matrix Scientific

009097-5G 009097-25G
101796-332EA 80 USD
101796-332 101796-334
Perfluoro-3,6-dioxadecanoic acid ≥98%
Perfluoro-3,6-dioxadecanoic acid

Hazard: Corrosive

Formula: C₈HF₁₅O₄
MDL Number: MFCD03094421
CAS Number: 137780-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR