Pentafluorophenyl methacrylate ≥97%

Supplier: Matrix Scientific

009082-1G 009082-5G
101796-294EA 121.54 USD
101796-294 101796-296
Pentafluorophenyl methacrylate ≥97%
Pentafluorophenyl methacrylate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₅F₅O₂
Boiling Pt: 56°C/3 Torr
MDL Number: MFCD00042332
CAS Number: 13642-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR