1H,1H,8H,8H-Octafluoro-3,6-dioxaoctane-1,8-diol ≥98%

Supplier: Matrix Scientific

Synonyms: 2,2'-((Perfluoroethane-1,2-diyl)bis(oxy))bis(2,2-difluoroethanol)

009072-5G 009072-25G
101796-276EA 164.62 USD
101796-276 101796-278
1H,1H,8H,8H-Octafluoro-3,6-dioxaoctane-1,8-diol ≥98%
1H,1H,8H,8H-Octafluoro-3,6-dioxaoctane-1,8-diol

Hazard: Irritant

Formula: C₆H₆F₈O₄
MW: 294.1 g/mol
MDL Number: MFCD03094435
CAS Number: 129301-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR