Methylisonicotinate-N-oxide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Methoxycarbonyl)pyridine 1-oxide

009063-5G 009063-25G
101796-246EA 60 USD
101796-246 101796-248
Methylisonicotinate-N-oxide ≥95%
Methylisonicotinate-N-oxide

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD00128856
CAS Number: 3783-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR