1H,1H-Nonafluoro-3,6-dioxaheptan-1-ol ≥98%

Supplier: Matrix Scientific

009051-5G 009051-25G
101796-206EA 135.38 USD
101796-206 101796-208
1H,1H-Nonafluoro-3,6-dioxaheptan-1-ol ≥98%
1H,1H-Nonafluoro-3,6-dioxaheptan-1-ol

Hazard: Irritant

Formula: C₅H₃F₉O₃
MDL Number: MFCD03094428
CAS Number: 330562-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR