N-Morpholinium trifluoromethanesulfonate
Supplier: Matrix Scientific
009044-5G
009044-25G
101796-188EA
223.08
USD
101796-188
101796-190
N-Morpholinium trifluoromethanesulfonate
N-Morpholinium trifluoromethanesulfonate
Hazard: Irritant
Formula:
C₅H₁₀F₃NO₄S MW: 237.2 g/mol |
MDL Number:
MFCD03094347 CAS Number: 77534-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...