N-Morpholinium nonafluorobutanesulfonate

Supplier: Matrix Scientific

009043-5G 009043-25G
101796-184EA 144.62 USD
101796-184 101796-186
N-Morpholinium nonafluorobutanesulfonate
N-Morpholinium nonafluorobutanesulfonate

Hazard: Irritant

Formula: C₈H₁₀F₉NO₄S
MW: 387.22 g/mol
MDL Number: MFCD03094228
CAS Number: 503155-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR