3,4,5,6-Tetrafluoro-N-methylphthalimide ≥97%

Supplier: Matrix Scientific

Synonyms: 4,5,6,7-Tetrafluoro-2-methylisoindoline-1,3-dione

009037-5G 009037-25G
101796-176EA 126.15 USD
101796-176 101796-178
3,4,5,6-Tetrafluoro-N-methylphthalimide ≥97%
3,4,5,6-Tetrafluoro-N-methylphthalimide

Hazard: Irritant

Formula: C₉H₃F₄NO₂
MW: 233.12 g/mol
MDL Number: MFCD01631699
CAS Number: 33795-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR