N-Methyl iodofluoroacetamide ≥98%

Supplier: Matrix Scientific

009030-1G 009030-5G
101796-154EA 78.46 USD
101796-154 101796-156
N-Methyl iodofluoroacetamide ≥98%
N-Methyl iodofluoroacetamide

Hazard: Irritant

Formula: C₃H₅FINO
MDL Number: MFCD04038321
CAS Number: 883499-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR