1-Iodo-1H,1H-perfluorooctane ≥97%

Supplier: Matrix Scientific

009013-1G 009013-5G
101796-114EA 112.31 USD
101796-114 101796-116
1-Iodo-1H,1H-perfluorooctane ≥97%
1-Iodo-1H,1H-perfluorooctane

Hazard: Irritant

Formula: C₈H₂F₁₅I
MDL Number: MFCD04038315
CAS Number: 10258-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR