1-Iodo-1H,1H-nonafluoropentane ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Iodo-1H,1H-perfluoropentane

009008-1G 009008-5G
101796-094EA 153.85 USD
101796-094 101796-096
1-Iodo-1H,1H-nonafluoropentane ≥97%
1-Iodo-1H,1H-nonafluoropentane

Hazard: Irritant

Formula: C₅H₂F₉I
MW: 359.96 g/mol
MDL Number: MFCD00233025
CAS Number: 2253-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR