2-(Iodomethyl)-1,1,1,3,3,3-hexafluoropropane ≥97%

Supplier: Matrix Scientific

009006-1G 009006-5G
101796-086EA 73.85 USD
101796-086 101796-088
2-(Iodomethyl)-1,1,1,3,3,3-hexafluoropropane ≥97%
2-(Iodomethyl)-1,1,1,3,3,3-hexafluoropropane

Hazard: Irritant

Formula: C₄H₃F₆I
MDL Number: MFCD04038313
CAS Number: 883449-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR