1,1,2,3,3,3-Hexafluoropropyl trichloromethyl ether ≥98%

Supplier: Matrix Scientific

008998-1G 008998-5G
101796-056EA 55.38 USD
101796-056 101796-058
1,1,2,3,3,3-Hexafluoropropyl trichloromethyl ether ≥98%
1,1,2,3,3,3-Hexafluoropropyl trichloromethyl ether

Hazard: Irritant

Formula: C₄HCl₃F₆O
MDL Number: MFCD03094226
CAS Number: 56860-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR