1H,1H-Heptafluorobutylnonafluorobutanesulfonate ≥97%

Supplier: Matrix Scientific

008977-1G 008977-5G
101795-994EA 76.92 USD
101795-994 101795-996
1H,1H-Heptafluorobutylnonafluorobutanesulfonate ≥97%
1H,1H-Heptafluorobutylnonafluorobutanesulfonate

Hazard: Toxic

Formula: C₈H₂F₁₆O₃S
MW: 482.14 g/mol
MDL Number: MFCD04038309
CAS Number: 883499-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR