4-(4-Fluorophenyl)-1-butene ≥97%

Supplier: Matrix Scientific

008963-1G 008963-5G
101795-952EA 55.38 USD
101795-952 101795-954
4-(4-Fluorophenyl)-1-butene ≥97%
4-(4-Fluorophenyl)-1-butene

Hazard: Flammable

Formula: C₁₀H₁₁F
MW: 150.193 g/mol
Boiling Pt: 72°C/9mmHg
Density: 0.97
MDL Number: MFCD00671360
CAS Number: 2248-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR