2,2-Difluoroethyl-p-toluenesulfonate ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluoroethyl 4-methylbenzenesulphonate

008921-1G 008921-5G
101795-824EA 64.62 USD
101795-824 101795-826
2,2-Difluoroethyl-p-toluenesulfonate ≥95%
2,2-Difluoroethyl-p-toluenesulfonate

Hazard: Irritant

Formula: C₉H₁₀F₂O₃S
MW: 236.24 g/mol
MDL Number: MFCD04038284
CAS Number: 135206-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR