3,3-Dichloro-1,1,1-trifluoroacetone trihydrate ≥99%

Supplier: Matrix Scientific

008909-5G 008909-25G
101795-792EA 80 USD
101795-792 101795-794
3,3-Dichloro-1,1,1-trifluoroacetone trihydrate ≥99%
3,3-Dichloro-1,1,1-trifluoroacetone trihydrate

Hazard: Irritant/Irritant-Harmful

Formula: C₃H₇Cl₂F₃O₄
MW: 234.99 g/mol
Boiling Pt: 75-76°C
MDL Number: MFCD04038282
CAS Number: 126266-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR