1,4-Dibromohexafluoro-2-butene ≥97%

Supplier: Matrix Scientific

008889-1G 008889-5G
101795-724EA 61.54 USD
101795-724 101795-726
1,4-Dibromohexafluoro-2-butene ≥97%
1,4-Dibromohexafluoro-2-butene

Hazard: Irritant/Refrigerate

Formula: C₄Br₂F₆
MW: 321.85 g/mol
MDL Number: MFCD04038279
CAS Number: 360-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR