3-Chloro-2-iodo-1,1,1-trifluoropropane ≥97%

Supplier: Matrix Scientific

008876-1G 008876-5G
101795-692EA 46.15 USD
101795-692 101795-694
3-Chloro-2-iodo-1,1,1-trifluoropropane ≥97%
3-Chloro-2-iodo-1,1,1-trifluoropropane

Hazard: Irritant

Formula: C₃H₃ClF₃I
MDL Number: MFCD01862016
CAS Number: 113402-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR