tert-Butyl pentafluoropropionate ≥98%

Supplier: Matrix Scientific

008864-1G 008864-5G
101795-656EA 63.08 USD
101795-656 101795-658
tert-Butyl pentafluoropropionate ≥98%
tert-Butyl pentafluoropropionate

Hazard: Irritant

Formula: C₇H₉F₅O₂
MW: 220.14 g/mol
MDL Number: MFCD00464380
CAS Number: 55258-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR