3-Bromo-1,1,1-trifluoroacetone oxime ≥97%

Supplier: Matrix Scientific

008855-1G 008855-5G
101795-628EA 121.54 USD
101795-628 101795-630
3-Bromo-1,1,1-trifluoroacetone oxime ≥97%
3-Bromo-1,1,1-trifluoroacetone oxime

Hazard: Irritant

Formula: C₃H₃BrF₃NO
MDL Number: MFCD04038272
CAS Number: 117341-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR