(1-Bromo)perfluoro-2,5,8-trioxadodecane ≥98%

Supplier: Matrix Scientific

Synonyms: Perfluoro-2,5-8-trioxadodecyl bromide

008848-5G 008848-25G
101795-600EA 153.85 USD
101795-600 101795-602
(1-Bromo)perfluoro-2,5,8-trioxadodecane ≥98%
(1-Bromo)perfluoro-2,5,8-trioxadodecane

Hazard: Irritant

Formula: C₉BrF₁₉O₃
MW: 596.98 g/mol
MDL Number: MFCD03094130
CAS Number: 330562-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR