(1-Bromo)perfluoro-2,5-dioxanonane ≥98%

Supplier: Matrix Scientific

008847-5G 008847-25G
101795-596EA 89.23 USD
101795-596 101795-598
(1-Bromo)perfluoro-2,5-dioxanonane ≥98%
(1-Bromo)perfluoro-2,5-dioxanonane

Hazard: Irritant

Formula: C₇BrF₁₅O₂
MDL Number: MFCD03094416
CAS Number: 330562-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR