2-Bromo-3-chloro-2-(trifluoromethyl)-1,1,1-trifluoropropane ≥97%

Supplier: Matrix Scientific

008827-1G 008827-5G
101795-544EA 63.08 USD
101795-544 101795-546
2-Bromo-3-chloro-2-(trifluoromethyl)-1,1,1-trifluoropropane ≥97%
2-Bromo-3-chloro-2-(trifluoromethyl)-1,1,1-trifluoropropane

Hazard: Irritant

Formula: C₄H₂BrClF₆
MDL Number: MFCD04038265
CAS Number: 883498-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR