1-Bromo-4-isobutylbenzene ≥98%

Supplier: Matrix Scientific

008824-25G 008824-100G
101795-534EA 110.77 USD
101795-534 101795-536
1-Bromo-4-isobutylbenzene ≥98%
1-Bromo-4-isobutylbenzene

Hazard: Irritant

Formula: C₁₀H₁₃Br
Boiling Pt: 97…98 °C (5 Torr)
Density: 1.2609
MDL Number: MFCD03425886
CAS Number: 2051-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR