Allyl-2,2,3,3,3-pentafluoropropyl ether ≥97%

Supplier: Matrix Scientific

008793-1G 008793-5G
101795-470EA 58.46 USD
101795-470 101795-472
Allyl-2,2,3,3,3-pentafluoropropyl ether ≥97%
Allyl-2,2,3,3,3-pentafluoropropyl ether

Hazard: Flammable/Irritant

Formula: C₆H₇F₅O
Boiling Pt: 85…86 °C
Flash Pt: 14 °C
MDL Number: MFCD02093320
CAS Number: 186907-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR