3-Nitro-4-(trifluoromethoxy)aniline ≥98%

Supplier: Matrix Scientific

008767-5G 008767-25G 008767-100G
101795-402EA 27.69 USD
101795-402 101795-404 101795-406
3-Nitro-4-(trifluoromethoxy)aniline ≥98%
3-Nitro-4-(trifluoromethoxy)aniline

Hazard: Irritant-Harmful

Formula: C₇H₅F₃N₂O₃
Melting Pt: 90…92 °C
MDL Number: MFCD03840207
CAS Number: 2822-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR