1-Chloro-3-(trifluoromethoxy)benzene ≥98%
Supplier: Matrix Scientific
008764-5G
008764-25G
008764-100G
101795-396EA
29.23
USD
101795-396
101795-398
101795-400
1-Chloro-3-(trifluoromethoxy)benzene ≥98%
1-Chloro-3-(trifluoromethoxy)benzene
Hazard: Irritant
Formula:
C₇H₄ClF₃O Boiling Pt: 142…143 °C Density: 1.369 |
MDL Number:
MFCD00276971 CAS Number: 772-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...