1-Chloro-2-(trifluoromethoxy)benzene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethoxy)chlorobenzene

008763-25G 008763-100G
101795-392EA 44.62 USD
101795-392 101795-394
1-Chloro-2-(trifluoromethoxy)benzene ≥98%
1-Chloro-2-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₇H₄ClF₃O
MW: 196.5543 g/mol
Boiling Pt: 142-143°C
Density: 1.369
MDL Number: MFCD03840206
CAS Number: 450-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR