3,3',5,5'-Tetramethylbiphenyl ≥95%

Supplier: Matrix Scientific

Synonyms: 3,3',5,5'-Tetramethyl-1,1'-biphenyl

067235-1G 067235-5G
101933-604EA 95.38 USD
101933-604 101933-606
3,3',5,5'-Tetramethylbiphenyl ≥95%
3,3',5,5'-Tetramethylbiphenyl

Hazard: Irritant

Formula: C₁₆H₁₈
MW: 210.32 g/mol
MDL Number: MFCD00051910
CAS Number: 25570-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR