Methyl-7-cyclopropyl-1-ethyl-2-mercapto-4-oxo-1,4-dihydropyrido[2,3-d]pyrimidine-5-carboxylate

Supplier: Matrix Scientific

063807-500MG
101926-418EA 364.62 USD
101926-418
Methyl-7-cyclopropyl-1-ethyl-2-mercapto-4-oxo-1,4-dihydropyrido[2,3-d]pyrimidine-5-carboxylate
Methyl-7-cyclopropyl-1-ethyl-2-mercapto-4-oxo-1,4-dihydropyrido[2,3-d]pyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₅N₃O₃S
MDL Number: MFCD09701475
CAS Number: 933949-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR