4,5-Dichloro-3(2H)-pyridazinone ≥95%
Supplier: Matrix Scientific
Synonyms:
4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one
063427-25G
063427-100G
101925-694EA
67.69
USD
101925-694
102833-332
4,5-Dichloro-3(2H)-pyridazinone ≥95%
4,5-Dichloro-3(2H)-pyridazinone
Hazard: Irritant
Formula:
C₄H₂Cl₂N₂O MW: 164.98 g/mol Melting Pt: 200-204°C |
MDL Number:
MFCD00051504 CAS Number: 932-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...