2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥95%

Supplier: Matrix Scientific

063415-100G 063415-1KG
101925-666EA 56.92 USD
101925-666 101925-668
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥95%
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)

Hazard: Irritant/Refrigerate

Formula: C₆H₈O₄
Melting Pt: 92-96°C
MDL Number: MFCD00006638
CAS Number: 2033-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR