6-Chloro-3-hydroxypyridazine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-chloropyridazin-3-ol, 6-Chloro-3(2H)-pyridazinone

063345-5G 063345-25G
101925-582EA 15.38 USD
101925-582 101925-584
6-Chloro-3-hydroxypyridazine ≥95%
6-Chloro-3-hydroxypyridazine

Hazard: Irritant

Formula: C₄H₃ClN₂O
MW: 130.53 g/mol
MDL Number: MFCD00160460
CAS Number: 19064-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR