Furan-2,5-dicarboxylic acid

Supplier: Matrix Scientific

Synonyms: 2,5-Furandicarboxylic acid

063028-5G 063028-25G
101925-000EA 36.92 USD
101925-000 101925-002
Furan-2,5-dicarboxylic acid
Furan-2,5-dicarboxylic acid

Hazard: Irritant

Formula: C₆H₄O₅
MW: 156.09 g/mol
Melting Pt: 310°C
MDL Number: MFCD00016582
CAS Number: 3238-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR