4-Chloro-5-fluoro-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde

Supplier: Matrix Scientific

059339-250MG 059339-1G
101917-952EA 606.15 USD
101917-952 101917-954
4-Chloro-5-fluoro-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde
4-Chloro-5-fluoro-1H-pyrrolo[2,3-b]pyridine-6-carbaldehyde

Hazard: Irritant

Formula: C₈H₄ClFN₂O
MDL Number: MFCD17171309
CAS Number: 1246088-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR