2-Amino-5-methylnicotinic acid

Supplier: Matrix Scientific

059338-250MG 059338-1G
101917-948EA 538.46 USD
101917-948 101917-950
2-Amino-5-methylnicotinic acid
2-Amino-5-methylnicotinic acid

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD07375376
CAS Number: 532440-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR