2,5-Difluoro-4-iodopyridine ≥97%

Supplier: Matrix Scientific

059330-1G 059330-5G
101917-934EA 44.62 USD
101917-934 102551-552
2,5-Difluoro-4-iodopyridine ≥97%
2,5-Difluoro-4-iodopyridine

Hazard: Irritant

Formula: C₅H₂F₂IN
MDL Number: MFCD11040606
CAS Number: 1017793-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR