3-Fluoro-5-methoxypyridine ≥98%

Supplier: Matrix Scientific

059326-250MG 059326-1G
101917-922EA 112.31 USD
101917-922 101917-924
3-Fluoro-5-methoxypyridine ≥98%
3-Fluoro-5-methoxypyridine

Hazard: Irritant

Formula: C₆H₆FNO
MDL Number: MFCD13185606
CAS Number: 1060801-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR