2-Fluoro-3-(furan-2-yl)-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

059307-500MG 059307-1G
101917-842EA 401.54 USD
101917-842 101917-844
2-Fluoro-3-(furan-2-yl)-3-oxopropanenitrile ≥95%
2-Fluoro-3-(furan-2-yl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₇H₄FNO₂
MDL Number: MFCD18426520
CAS Number: 1263063-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR