Ethyl-4-bromo-3-oxopentanoate ≥90%

Supplier: Matrix Scientific

059304-500MG 059304-1G 059304-5G
101917-828EA 295.38 USD
101917-828 101917-830 101917-832
Ethyl-4-bromo-3-oxopentanoate ≥90%
Ethyl-4-bromo-3-oxopentanoate

Hazard: Irritant

Formula: C₇H₁₁BrO₃
MDL Number: MFCD17014835
CAS Number: 36187-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR