5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride ≥95%
Supplier: Matrix Scientific
059276-1G
059276-5G
101917-756EA
292.31
USD
101917-756
101917-758
5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride ≥95%
5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride
Hazard: Irritant
Formula:
C₇H₇ClF₃N₃ |
MDL Number:
MFCD00203995 CAS Number: 175277-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...